7-Chloro-n-p-tolylthiazolo[5,4-d]pyrimidin-2-amine

Lieferant: Apollo Scientific

Synonyme: 7-Chloro-N-(p-tolyl)thiazolo[5,4-d]pyrimidin-2-amine

OR322974-250MG
APOSOR322974-250MGEA 78.3 EUR
APOSOR322974-250MG
7-Chloro-n-p-tolylthiazolo[5,4-d]pyrimidin-2-amine
7-Chloro-n-p-tolylthiazolo[5,4-d]pyrimidin-2-amine
Formel: C₁₂H₉ClN₄S
Molecular Weight: 276,745 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08437191
CAS-Nummer: 871266-80-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR