2-[6-(Dimethylamino)pyridazin-3-yl]-2-phenylacetonitrile, tech

Lieferant: Apollo Scientific

OR32298-1G
APOSOR32298-1GEA 1570 EUR
APOSOR32298-1G
2-[6-(Dimethylamino)pyridazin-3-yl]-2-phenylacetonitrile, tech
2-[6-(Dimethylamino)pyridazin-3-yl]-2-phenylacetonitrile
Formel: C₁₄H₁₄N₄
Molecular Weight: 238,288 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00243407
CAS-Nummer: 303997-53-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR