Ethyl 1-(4-chlorophenyl)-5-methyl-1H-1,2,4-triazole-3-carboxylate, tech

Lieferant: Apollo Scientific

OR32299-1G
APOSOR32299-1GEA 1570 EUR
APOSOR32299-1G
Ethyl 1-(4-chlorophenyl)-5-methyl-1H-1,2,4-triazole-3-carboxylate, tech
Ethyl 1-(4-chlorophenyl)-5-methyl-1H-1,2,4-triazole-3-carboxylate
Formel: C₁₂H₁₂ClN₃O₂
Molecular Weight: 265,696 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00173065
CAS-Nummer: 91396-27-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR