4-Chloro-N-phenylbenzene-1-sulphonamide, tech
Lieferant: Apollo Scientific
OR32302-1G
APOSOR32302-1GEA
780
EUR
APOSOR32302-1G
4-Chloro-N-phenylbenzene-1-sulphonamide, tech
4-Chloro-N-phenylbenzene-1-sulphonamide
Formel:
C₁₂H₁₀ClNO₂S Molecular Weight: 267,731 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00443139 CAS-Nummer: 7454-47-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...