3-(2-Chlorophenoxy)butan-2-one, tech
Lieferant: Apollo Scientific
OR32304-1G
APOSOR32304-1GEA
1570
EUR
APOSOR32304-1G
3-(2-Chlorophenoxy)butan-2-one, tech
3-(2-Chlorophenoxy)butan-2-one
Formel:
C₁₀H₁₁ClO₂ Molecular Weight: 198,646 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00243571 CAS-Nummer: 3757-62-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...