3-(3-Methoxyphenoxy)butan-2-one, tech

Lieferant: Apollo Scientific

OR32305-1G
APOSOR32305-1GEA 1570 EUR
APOSOR32305-1G
3-(3-Methoxyphenoxy)butan-2-one, tech
3-(3-Methoxyphenoxy)butan-2-one
Formel: C₁₁H₁₄O₃
Molecular Weight: 194,227 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00243572
CAS-Nummer: 93351-44-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR