3-(3-Methylphenoxy)butan-2-one, tech

Lieferant: Apollo Scientific

OR32306-1G
APOSOR32306-1GEA 1570 EUR
APOSOR32306-1G
3-(3-Methylphenoxy)butan-2-one, tech
3-(3-Methylphenoxy)butan-2-one
Formel: C₁₁H₁₄O₂
Molecular Weight: 178,228 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00243573
CAS-Nummer: 6560-02-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR