3-(4-Methylphenoxy)butan-2-one, tech

Lieferant: Apollo Scientific

OR32310-1G
APOSOR32310-1GEA 1570 EUR
APOSOR32310-1G
3-(4-Methylphenoxy)butan-2-one, tech
3-(4-Methylphenoxy)butan-2-one
Formel: C₁₁H₁₄O₂
Molecular Weight: 178,228 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00243577
CAS-Nummer: 91969-89-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR