4-{[(4E)-5-(Dimethylamino)-3-oxopent-4-en-2-yl]oxy}benzonitrile, tech

Lieferant: Apollo Scientific

OR32312-1G
APOSOR32312-1GEA 1570 EUR
APOSOR32312-1G
4-{[(4E)-5-(Dimethylamino)-3-oxopent-4-en-2-yl]oxy}benzonitrile, tech
4-{[(4E)-5-(Dimethylamino)-3-oxopent-4-en-2-yl]oxy}benzonitrile
Formel: C₁₄H₁₆N₂O₂
Molecular Weight: 244,289 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00243583
CAS-Nummer: 306979-63-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR