3-(2,6-Dimethylphenoxy)butan-2-one, tech
Lieferant: Apollo Scientific
OR32313-1G
APOSOR32313-1GEA
1570
EUR
APOSOR32313-1G
3-(2,6-Dimethylphenoxy)butan-2-one, tech
3-(2,6-Dimethylphenoxy)butan-2-one
Formel:
C₁₂H₁₆O₂ Molecular Weight: 192,254 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00243586 CAS-Nummer: 25826-38-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...