5-Acetyl-2-[benzyl(methyl)amino]-6-methylpyridine-3-carbonitrile, tech

Lieferant: Apollo Scientific

OR32314-1G
APOSOR32314-1GEA 1570 EUR
APOSOR32314-1G
5-Acetyl-2-[benzyl(methyl)amino]-6-methylpyridine-3-carbonitrile, tech
5-Acetyl-2-[benzyl(methyl)amino]-6-methylpyridine-3-carbonitrile
Formel: C₁₇H₁₇N₃O
Molecular Weight: 279,336 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00793613
CAS-Nummer: 306979-97-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR