(3Z)-3-[(Dimethylamino)methylidene]-6-methyl-3,4-dihydro-2H-pyran-2,4-dione, tech

Lieferant: Apollo Scientific

OR32316-1G
APOSOR32316-1GEA 1570 EUR
APOSOR32316-1G
(3Z)-3-[(Dimethylamino)methylidene]-6-methyl-3,4-dihydro-2H-pyran-2,4-dione, tech
(3Z)-3-[(Dimethylamino)methylidene]-6-methyl-3,4-dihydro-2H-pyran-2,4-dione
Formel: C₉H₁₁NO₃
Molecular Weight: 181,189 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00794119
CAS-Nummer: 181277-21-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR