(3Z)-3-{[(2,4-Dimethylphenyl)amino]methylidene}-6-methyl-3,4-dihydro-2H-pyran-2,4-dione, tech

Lieferant: Apollo Scientific

OR32317-1G
APOSOR32317-1GEA 1570 EUR
APOSOR32317-1G
(3Z)-3-{[(2,4-Dimethylphenyl)amino]methylidene}-6-methyl-3,4-dihydro-2H-pyran-2,4-dione, tech
(3Z)-3-{[(2,4-Dimethylphenyl)amino]methylidene}-6-methyl-3,4-dihydro-2H-pyran-2,4-dione
Formel: C₁₅H₁₅NO₃
Molecular Weight: 257,284 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00794131
CAS-Nummer: 477859-27-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR