(2S,4R)-Methyl 4-hydroxy-1-methylpyrrolidine-2-carboxylate

Lieferant: Apollo Scientific

Synonyme: (R)-1-Methyl-4-hydroxy-L-proline methyl ester

OR323173-250MG
APOSOR323173-250MGEA 59.6 EUR
APOSOR323173-250MG
(2S,4R)-Methyl 4-hydroxy-1-methylpyrrolidine-2-carboxylate
(2S,4R)-Methyl 4-hydroxy-1-methylpyrrolidine-2-carboxylate
Formel: C₇H₁₃NO₃
Molecular Weight: 159,183 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18071015
CAS-Nummer: 13135-69-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR