4-((Dimethylamino)methyl)benzonitrile

Lieferant: Apollo Scientific

OR323176-1G
APOSOR323176-1GEA 30.6 EUR
APOSOR323176-1G
4-((Dimethylamino)methyl)benzonitrile
4-((Dimethylamino)methyl)benzonitrile
Formel: C₁₀H₁₂N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00029578
CAS-Nummer: 35525-86-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR