(3Z)-6-Methyl-3-({[4-(propan-2-yl)phenyl]amino}methylidene)-3,4-dihydro-2H-pyran-2,4-dione, tech

Lieferant: Apollo Scientific

OR32318-1G
APOSOR32318-1GEA 1570 EUR
APOSOR32318-1G
(3Z)-6-Methyl-3-({[4-(propan-2-yl)phenyl]amino}methylidene)-3,4-dihydro-2H-pyran-2,4-dione, tech
(3Z)-6-Methyl-3-({[4-(propan-2-yl)phenyl]amino}methylidene)-3,4-dihydro-2H-pyran-2,4-dione
Formel: C₁₆H₁₇NO₃
Molecular Weight: 271,311 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00794132
CAS-Nummer: 477859-28-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR