5-(Hydroxymethyl)pyridin-3-ol

Lieferant: Apollo Scientific

OR323193-250MG
APOSOR323193-250MGEA 226 EUR
APOSOR323193-250MG
5-(Hydroxymethyl)pyridin-3-ol
5-(Hydroxymethyl)pyridin-3-ol
Formel: C₆H₇NO₂
Molecular Weight: 125,125 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12546435
CAS-Nummer: 51035-70-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR