Isopropyl piperazine-1-carboxylate

Lieferant: Apollo Scientific

OR323204-1G
APOSOR323204-1GEA 517 EUR
APOSOR323204-1G
Isopropyl piperazine-1-carboxylate
Isopropyl piperazine-1-carboxylate
Formel: C₈H₁₆N₂O₂
Molecular Weight: 172,225 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14585267
CAS-Nummer: 37038-26-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR