4-Acetyl-4-pentylcyclohexan-1-one, tech

Lieferant: Apollo Scientific

OR32321-1G
APOSOR32321-1GEA 1570 EUR
APOSOR32321-1G
4-Acetyl-4-pentylcyclohexan-1-one, tech
4-Acetyl-4-pentylcyclohexan-1-one
Formel: C₁₃H₂₂O₂
Molecular Weight: 210.313 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02082370
CAS-Nummer: 76921-65-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR