5-Bromo-4-pyrimidinecarboxylic acid 95%

Lieferant: Apollo Scientific

OR323224-1G
APOSOR323224-1GEA 85.7 EUR
APOSOR323224-1G
5-Bromo-4-pyrimidinecarboxylic acid 95%
5-Bromo-4-pyrimidinecarboxylic acid
Formel: C₅H₃BrN₂O₂
Molecular Weight: 202,9900 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09754104
CAS-Nummer: 64224-60-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR