5,6-Dibromocyclohex-2-ene-1,4-dione, tech

Lieferant: Apollo Scientific

OR32323-1G
APOSOR32323-1GEA 1570 EUR
APOSOR32323-1G
5,6-Dibromocyclohex-2-ene-1,4-dione, tech
5,6-Dibromocyclohex-2-ene-1,4-dione
Formel: C₆H₄Br₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01568777
CAS-Nummer: 5273-61-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR