1-(4-Nitrophenylmethyl)piperidine

Lieferant: Apollo Scientific

Synonyme: 1-(4-Nitrobenzyl)piperidine

OR323231-1G
APOSOR323231-1GEA 60.9 EUR
APOSOR323231-1G
1-(4-Nitrophenylmethyl)piperidine
1-(4-Nitrophenylmethyl)piperidine
Formel: C₁₂H₁₆N₂O₂
Molecular Weight: 220,268 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01453278
CAS-Nummer: 59507-44-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR