(2R,3R)-3-((S)-1-(tert-Butoxycarbonyl)pyrrolidin-2-yl)-3-methoxy-2-methylpropanoic acid

Lieferant: Apollo Scientific

Synonyme: N-Boc-dolaproine

OR323233-250MG
APOSOR323233-250MGEA 102 EUR
APOSOR323233-250MG
(2R,3R)-3-((S)-1-(tert-Butoxycarbonyl)pyrrolidin-2-yl)-3-methoxy-2-methylpropanoic acid
(2R,3R)-3-((S)-1-(tert-Butoxycarbonyl)pyrrolidin-2-yl)-3-methoxy-2-methylpropanoic acid
Formel: C₁₄H₂₅NO₅
Molecular Weight: 287,352 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18157735
CAS-Nummer: 120205-50-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR