3-(3,5-Dibromo-1H-1,2,4-triazol-1-yl)butan-2-one, tech

Lieferant: Apollo Scientific

OR32324-1G
APOSOR32324-1GEA 1570 EUR
APOSOR32324-1G
3-(3,5-Dibromo-1H-1,2,4-triazol-1-yl)butan-2-one, tech
3-(3,5-Dibromo-1H-1,2,4-triazol-1-yl)butan-2-one
Formel: C₆H₇Br₂N₃O
Molecular Weight: 296,947 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01568885
CAS-Nummer: 320424-38-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR