Methyl-1-(cyanomethyl)-4-piperidinecarboxylate 95%
Lieferant: Apollo Scientific
OR32325-1G
OR32325-5G
APOSOR32325-1GEA
336
EUR
APOSOR32325-1G
APOSOR32325-5G
Methyl-1-(cyanomethyl)-4-piperidinecarboxylate 95%
Methyl-1-(cyanomethyl)-4-piperidinecarboxylate
Formel:
C₉H₁₄N₂O₂ Molecular Weight: 182.22 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01568888 CAS-Nummer: 320424-43-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...