(Z)-(Ethyl N-[(Z)-benzoyl]ethanecarbohydrazonate), tech

Lieferant: Apollo Scientific

OR32326-1G
APOSOR32326-1GEA 1570 EUR
APOSOR32326-1G
(Z)-(Ethyl N-[(Z)-benzoyl]ethanecarbohydrazonate), tech
(Z)-(Ethyl N-[(Z)-benzoyl]ethanecarbohydrazonate)
Formel: C₁₁H₁₄N₂O₂
Molecular Weight: 206,241 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02082701
CAS-Nummer: 85936-50-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR