1,3-Diethylharnstoff 97%

Lieferant: Apollo Scientific

Synonyme: N,N'-Diethylurea

OR323264-100G
APOSOR323264-100GEA 104 EUR
APOSOR323264-100G
1,3-Diethylharnstoff 97%
1,3-Diethylharnstoff
Formel: C₅H₁₂N₂O
Molecular Weight: 116.16 g/mol
Schmelzpunkt: 109…111 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00009028
CAS-Nummer: 623-76-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR