(2-Aminoethyl)phosphonic acid 97%
Lieferant: Apollo Scientific
Synonyme:
2-AEP, 2-Aminoethylphosphonic acid
OR323272-1G
APOSOR323272-1GEA
82.8
EUR
APOSOR323272-1G
(2-Aminoethyl)phosphonic acid 97%
(2-Aminoethyl)phosphonic acid
Formel:
C₂H₈NO₃P Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008182 CAS-Nummer: 2041-14-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...