5-Methoxy-N-methyl-2-phenylpyrimidin-4-amine, tech

Lieferant: Apollo Scientific

OR32328-1G
APOSOR32328-1GEA 1570 EUR
APOSOR32328-1G
5-Methoxy-N-methyl-2-phenylpyrimidin-4-amine, tech
5-Methoxy-N-methyl-2-phenylpyrimidin-4-amine
Formel: C₁₂H₁₃N₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD02102335
CAS-Nummer: 477863-75-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR