5-Methoxy-2-phenyl-N-(prop-2-en-1-yl)pyrimidin-4-amine, tech

Lieferant: Apollo Scientific

OR32329-1G
APOSOR32329-1GEA 1570 EUR
APOSOR32329-1G
5-Methoxy-2-phenyl-N-(prop-2-en-1-yl)pyrimidin-4-amine, tech
5-Methoxy-2-phenyl-N-(prop-2-en-1-yl)pyrimidin-4-amine
Formel: C₁₄H₁₅N₃O
Molecular Weight: 241,288 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02102339
CAS-Nummer: 477863-87-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR