N-Boc-glycin (Boc-Gly-OH) 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: Boc-Gly-OH

OR323299-100G OR323299-1KG OR323299-500G
APOSOR323299-100GEA 21.9 EUR
APOSOR323299-100G APOSOR323299-1KG APOSOR323299-500G
N-Boc-glycin (Boc-Gly-OH) 97%
N-Boc-glycin (Boc-Gly-OH)
Formel: C₇H₁₃NO₄
Schmelzpunkt: 87…90 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00002690
CAS-Nummer: 4530-20-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR