(R)-(+)-2-Methyl-2-propanesulphinamide 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(R)-2-methylpropane-2-sulphinamide, R)-(+)-tert-Butyl sulphinamide, (R)-2-Methyl-2-propanesulphinamide, (R)-(+)-tert-Butylsulphinamide, (R)-(+)-t-Butylsulphinamide
OR3233-100G
OR3233-25G
OR3233-500G
APOSOR3233-100GEA
84.1
EUR
APOSOR3233-100G
APOSOR3233-25G
APOSOR3233-500G
(R)-(+)-2-Methyl-2-propanesulphinamide 98%
(R)-(+)-2-Methyl-2-propanesulphinamide
Formel:
C₄H₁₁NOS Schmelzpunkt: 103…107 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD05861479 CAS-Nummer: 196929-78-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...