1-(7-Methoxy-1-benzofuran-2-yl)ethan-1-ol, tech

Lieferant: Apollo Scientific

OR32330-1G
APOSOR32330-1GEA 1570 EUR
APOSOR32330-1G
1-(7-Methoxy-1-benzofuran-2-yl)ethan-1-ol, tech
1-(7-Methoxy-1-benzofuran-2-yl)ethan-1-ol
Formel: C₁₁H₁₂O₃
Molecular Weight: 192,211 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02082714
CAS-Nummer: 260786-00-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR