(3E)-3-[(Dimethylamino)methylidene]-3,4-dihydro-2H-1-benzopyran-4-one, tech

Lieferant: Apollo Scientific

OR32331-1G
APOSOR32331-1GEA 1570 EUR
APOSOR32331-1G
(3E)-3-[(Dimethylamino)methylidene]-3,4-dihydro-2H-1-benzopyran-4-one, tech
(3E)-3-[(Dimethylamino)methylidene]-3,4-dihydro-2H-1-benzopyran-4-one
Formel: C₁₂H₁₃NO₂
Molecular Weight: 203,237 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02082835
CAS-Nummer: 1340477-34-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR