2-(1-Hydroxy-3-phenylpropan-2-yl)-2,3-dihydro-1H-isoindol-1-one, tech

Lieferant: Apollo Scientific

OR32333-1G
APOSOR32333-1GEA 1570 EUR
APOSOR32333-1G
2-(1-Hydroxy-3-phenylpropan-2-yl)-2,3-dihydro-1H-isoindol-1-one, tech
2-(1-Hydroxy-3-phenylpropan-2-yl)-2,3-dihydro-1H-isoindol-1-one
Formel: C₁₇H₁₇NO₂
Molecular Weight: 267.322 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02082957
CAS-Nummer: 477864-32-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR