2,2-Dimethyl-4-(2-oxiranyl)tetrahydrofuro[3,4-d][1,3]dioxole 95%
Lieferant: Apollo Scientific
Synonyme:
2-{2,2-Dimethyl-tetrahydro-2H-furo[3,4-d][1,3]dioxol-4-yl}oxirane
OR32336-1G
APOSOR32336-1GEA
538
EUR
APOSOR32336-1G
2,2-Dimethyl-4-(2-oxiranyl)tetrahydrofuro[3,4-d][1,3]dioxole 95%
2,2-Dimethyl-4-(2-oxiranyl)tetrahydrofuro[3,4-d][1,3]dioxole
Formel:
C₉H₁₄O₄ Molecular Weight: 186,21 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03425721 CAS-Nummer: 226709-43-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...