6-(4-Chlorophenyl)-2-methylpyrazolo[1,5-a]pyrimidin-7-amine, tech
Lieferant: Apollo Scientific
OR32337-1G
APOSOR32337-1GEA
1570
EUR
APOSOR32337-1G
6-(4-Chlorophenyl)-2-methylpyrazolo[1,5-a]pyrimidin-7-amine, tech
6-(4-Chlorophenyl)-2-methylpyrazolo[1,5-a]pyrimidin-7-amine
Formel:
C₁₃H₁₁ClN₄ Molecular Weight: 258,706 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02661637 CAS-Nummer: 85841-04-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...