2-[(Dimethylamino)methylidene]-2,3-dihydro-1H-indene-1,3-dione, tech
Lieferant: Apollo Scientific
OR32339-1G
APOSOR32339-1GEA
1570
EUR
APOSOR32339-1G
2-[(Dimethylamino)methylidene]-2,3-dihydro-1H-indene-1,3-dione, tech
2-[(Dimethylamino)methylidene]-2,3-dihydro-1H-indene-1,3-dione
Formel:
C₁₂H₁₁NO₂ Molecular Weight: 201,221 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00557302 CAS-Nummer: 28752-91-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...