(2E)-3-(Dimethylamino)-1-[4-(1H-pyrrol-1-yl)phenyl]prop-2-en-1-one, tech
Lieferant: Apollo Scientific
OR32341-1G
APOSOR32341-1GEA
1570
EUR
APOSOR32341-1G
(2E)-3-(Dimethylamino)-1-[4-(1H-pyrrol-1-yl)phenyl]prop-2-en-1-one, tech
(2E)-3-(Dimethylamino)-1-[4-(1H-pyrrol-1-yl)phenyl]prop-2-en-1-one
Formel:
C₁₅H₁₆N₂O Molecular Weight: 240,3 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02661662 CAS-Nummer: 551930-66-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...