1-(4-Acetylphenyl)-1H-pyrrole-2-carbaldehyde, tech
Lieferant: Apollo Scientific
OR32342-1G
APOSOR32342-1GEA
1570
EUR
APOSOR32342-1G
1-(4-Acetylphenyl)-1H-pyrrole-2-carbaldehyde, tech
1-(4-Acetylphenyl)-1H-pyrrole-2-carbaldehyde
Formel:
C₁₃H₁₁NO₂ Molecular Weight: 213,232 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02661663 CAS-Nummer: 439094-81-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...