4-[4-(1H-Pyrrol-1-yl)phenyl]pyrimidin-2-amine, tech

Lieferant: Apollo Scientific

OR32343-1G
APOSOR32343-1GEA 1570 EUR
APOSOR32343-1G
4-[4-(1H-Pyrrol-1-yl)phenyl]pyrimidin-2-amine, tech
4-[4-(1H-Pyrrol-1-yl)phenyl]pyrimidin-2-amine
Formel: C₁₄H₁₂N₄
Molecular Weight: 236,272 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03102687
CAS-Nummer: 691870-09-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR