N-(2-Amino-5-chloro-4-methylphenyl)acetamide, tech

Lieferant: Apollo Scientific

OR32344-1G
APOSOR32344-1GEA 1570 EUR
APOSOR32344-1G
N-(2-Amino-5-chloro-4-methylphenyl)acetamide, tech
N-(2-Amino-5-chloro-4-methylphenyl)acetamide
Formel: C₉H₁₁ClN₂O
Molecular Weight: 198,649 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03011787
CAS-Nummer: 383147-42-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR