2-[4-(4-Chlorophenyl)-3-methyl-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl]acetonitrile, tech

Lieferant: Apollo Scientific

OR32348-1G
APOSOR32348-1GEA 1570 EUR
APOSOR32348-1G
2-[4-(4-Chlorophenyl)-3-methyl-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl]acetonitrile, tech
2-[4-(4-Chlorophenyl)-3-methyl-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl]acetonitrile
Formel: C₁₁H₉ClN₄O
Molecular Weight: 248.668 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03787637
CAS-Nummer: 860786-60-3

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR