2-[3-Methyl-4-(4-methylphenyl)-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl]acetonitrile, tech

Lieferant: Apollo Scientific

OR32349-1G
APOSOR32349-1GEA 1570 EUR
APOSOR32349-1G
2-[3-Methyl-4-(4-methylphenyl)-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl]acetonitrile, tech
2-[3-Methyl-4-(4-methylphenyl)-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl]acetonitrile
Formel: C₁₂H₁₂N₄O
Molecular Weight: 228,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03787670
CAS-Nummer: 860785-01-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR