2-Methyl-4,5-dihydro-1,3,4-benzoxadiazepin-5-one, tech

Lieferant: Apollo Scientific

OR32350-1G
APOSOR32350-1GEA 1570 EUR
APOSOR32350-1G
2-Methyl-4,5-dihydro-1,3,4-benzoxadiazepin-5-one, tech
2-Methyl-4,5-dihydro-1,3,4-benzoxadiazepin-5-one
Formel: C₉H₈N₂O₂
Molecular Weight: 176,172 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00268745
CAS-Nummer: 144760-62-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR