Ethyl 4-chloro-1-ethyl-3-methyl-1H-pyrazolo[3,4-b]pyridine-5-carboxylate 95%

Lieferant: Apollo Scientific

OR32351-1G
APOSOR32351-1GEA 874 EUR
APOSOR32351-1G
Ethyl 4-chloro-1-ethyl-3-methyl-1H-pyrazolo[3,4-b]pyridine-5-carboxylate 95%
Ethyl 4-chloro-1-ethyl-3-methyl-1H-pyrazolo[3,4-b]pyridine-5-carboxylate
Formel: C₁₂H₁₄ClN₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD04124526
CAS-Nummer: 37801-47-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR