2-Chloro-N-ethyl-N-phenylacetamide 95%

Lieferant: Apollo Scientific

OR32352-1G
APOSOR32352-1GEA 180 EUR
APOSOR32352-1G
2-Chloro-N-ethyl-N-phenylacetamide 95%
2-Chloro-N-ethyl-N-phenylacetamide
Formel: C₁₀H₁₂ClNO
Molecular Weight: 197.67 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00227876
CAS-Nummer: 39086-61-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR