Ethyl 1-[(6-chloropyridin-3-yl)methyl]piperidine-4-carboxylate, tech

Lieferant: Apollo Scientific

OR32357-1G
APOSOR32357-1GEA 1570 EUR
APOSOR32357-1G
Ethyl 1-[(6-chloropyridin-3-yl)methyl]piperidine-4-carboxylate, tech
Ethyl 1-[(6-chloropyridin-3-yl)methyl]piperidine-4-carboxylate
Formel: C₁₄H₁₉ClN₂O₂
Molecular Weight: 282,766 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04124605
CAS-Nummer: 860787-11-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR