3-[(4-Chlorophenyl)methyl]-5,5-dimethylimidazolidine-2,4-dione, tech

Lieferant: Apollo Scientific

OR32359-1G
APOSOR32359-1GEA 1570 EUR
APOSOR32359-1G
3-[(4-Chlorophenyl)methyl]-5,5-dimethylimidazolidine-2,4-dione, tech
3-[(4-Chlorophenyl)methyl]-5,5-dimethylimidazolidine-2,4-dione
Formel: C₁₂H₁₃ClN₂O₂
Molecular Weight: 252,697 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05975050
CAS-Nummer: 860787-34-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR