1-[4-(4-Chlorobenzoyl)piperidin-1-yl]ethan-1-one 95%

Lieferant: Apollo Scientific

OR32361-1G
APOSOR32361-1GEA 1120 EUR
APOSOR32361-1G
1-[4-(4-Chlorobenzoyl)piperidin-1-yl]ethan-1-one 95%
1-[4-(4-Chlorobenzoyl)piperidin-1-yl]ethan-1-one
Formel: C₁₄H₁₆ClNO₂
Molecular Weight: 265,735 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01566224
CAS-Nummer: 59084-15-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR